Assay Details
Target Gene Details
Entrez Gene ID: | 216033 |
Gene Name: | catenin (cadherin associated protein), alpha 3 |
Gene Aliases: |
4930429L08Rik, 4933408A16, Catna3, Vr22 |
Location: |
Chr.10:63430098-65005395 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctnna3 | NM_001164376.1 | 18 | 2691 | NP_001157848.1 |
NM_001164519.1 | 13 | 1850 | NP_001157991.1 | |
NM_177612.3 | 18 | 2619 | NP_808280.2 | |
AF344871.1 | 18 | 2691 | AAQ14965.1 | |
AK030166.1 | 18 | 2620 | BAC26817.1 | |
AK142745.1 | 18 | 2692 | BAE25182.1 | |
BC112409.1 | 18 | 2593 | ||
DQ380429.1 | 18 | 2619 | ||
DQ380430.1 | 13 | 1849 |
More Information
Set Membership: |
Intragenic Exonic |