Assay Details
Target Gene Details
Entrez Gene ID: | 103161 |
Gene Name: | apolipoprotein F |
Gene Aliases: |
AI255964 |
Location: |
Chr.10:128267997-128270151 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apof | NM_133997.2 | 2 | 963 | NP_598758.1 |
BC010815.1 | 2 | 225 | AAH10815.1 | |
BC022795.1 | 2 | 262 | AAH22795.1 | |
BC024769.1 | 2 | 250 | AAH24769.1 | |
BC025827.1 | 2 | 237 | AAH25827.1 |
More Information
Set Membership: |
Intragenic Exonic |