Assay Details
Target Gene Details
Entrez Gene ID: | 72774 |
Gene Name: | nei endonuclease VIII-like 1 (E. coli) |
Gene Aliases: |
2810450N13Rik, Nei1 |
Location: |
Chr.9:57142788-57148675 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Neil1 | NM_028347.2 | 1 | 72 | NP_082623.1 |
AB079069.1 | 1 | 72 | BAC06477.1 | |
AK013322.1 | 2 | 143 | BAB28790.1 | |
AK040802.1 | 2 | 129 | ||
AK155405.1 | 2 | 137 | BAE33246.1 | |
BC043297.1 | 2 | 236 | AAH43297.1 |
More Information
Set Membership: |
Intragenic Exonic |