Assay Details
Target Gene Details
Entrez Gene ID: | 72330 |
Gene Name: | kelch-like 40 |
Gene Aliases: |
2310024D23Rik, Kbtbd5 |
Location: |
Chr.9:121777101-121791002 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klhl40 | NM_028202.3 | 1 | 530 | NP_082478.1 |
AK009491.1 | 1 | 447 | BAB26321.1 | |
AK078669.1 | 1 | 443 | ||
AK160799.1 | 1 | 390 | BAE36021.1 | |
BC125576.1 | 1 | 530 | ||
BC137889.1 | 1 | 530 |
More Information
Set Membership: |
Intragenic Exonic |