Assay Details
Target Gene Details
Entrez Gene ID: | 69836 |
Gene Name: | phospholipase A2, group XIIB |
Gene Aliases: |
2010002E04Rik, Fksg71, Pla2g13, hlb218 |
Location: |
Chr.10:59403420-59421976 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g12b | NM_023530.2 | 4 | 980 | NP_076019.2 |
AK008035.1 | 4 | 1006 | ||
AY358033.1 | 4 | 1009 | AAQ92919.1 | |
BC021592.1 | 4 | 980 | AAH21592.1 |
More Information
Set Membership: |
Intragenic Exonic |