Assay Details
Target Gene Details
Entrez Gene ID: | 60406 |
Gene Name: | sin3 associated polypeptide |
Gene Aliases: |
30kDa |
Location: |
Chr.8:57482702-57487860 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sap30 | NM_021788.2 | 4 | 942 | NP_068560.1 |
AF075136.1 | 4 | 953 | AAC26007.1 | |
AK010928.1 | 4 | 943 | BAB27273.1 | |
AK088745.1 | 4 | 947 | BAC40543.1 | |
AV144112.1 | ||||
BC132081.1 | 4 | 925 | ||
BC132087.1 | 4 | 925 | ||
CO800093.1 | 1 | 259 |
More Information
Set Membership: |
Intragenic Exonic |