Assay Details
Target Gene Details
Entrez Gene ID: | 14182 |
Gene Name: | fibroblast growth factor receptor 1 |
Gene Aliases: |
AW208770, Eask, FLG, Fgfr-1, Flt-2, Hspy, MFR, bFGF-R-1, c-fgr |
Location: |
Chr.8:25513623-25575718 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgfr1 | NM_001079908.2 | 18 | 3375 | NP_001073377.1 |
NM_001079909.2 | 17 | 3114 | NP_001073378.1 | |
NM_010206.3 | 18 | 3381 | NP_034336.2 | |
AF176552.1 | 16 | 2412 | AAF05312.1 | |
AK028354.1 | 17 | 2554 | BAC25899.1 | |
AK034295.1 | 8 | 1201 | BAC28662.1 | |
AK049704.1 | 18 | 3341 | ||
AK167639.1 | 9 | 2678 | BAE39690.1 | |
BC010200.1 | 18 | 3274 | AAH10200.1 | |
BC033447.1 | 16 | 2377 | AAH33447.1 | |
M33760.1 | 16 | 2398 | AAA37622.1 |
More Information
Set Membership: |
Intragenic Exonic |