Assay Details
Target Gene Details
Entrez Gene ID: | 170786 |
Gene Name: | CD209a antigen |
Gene Aliases: |
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, SIGN-R1, SIGNR5 |
Location: |
Chr.8:3743395-3748984 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd209a | NM_133238.5 | 7 | 784 | NP_573501.1 |
AF373408.1 | 7 | 731 | AAL13234.1 | |
AF374470.1 | 6 | 650 | AAL27539.1 | |
AK054139.1 | 7 | 748 | BAC35667.1 | |
AK171639.1 | 7 | 728 | BAE42579.1 | |
AY049062.1 | 7 | 784 | AAK85825.1 | |
BC111095.1 | 6 | 832 | AAI11096.1 |
More Information
Set Membership: |
Intragenic Exonic |