Assay Details
Target Gene Details
Entrez Gene ID: | 80901 |
Gene Name: | chemokine (C-X-C motif) receptor 6 |
Gene Aliases: |
BB217514, BONZO, STRL33 |
Location: |
Chr.9:123806467-123811756 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcr6 | NM_030712.4 | 2 | 1589 | NP_109637.3 |
AF305709.1 | 2 | 1592 | AAG31284.1 | |
AK052901.1 | 2 | 1587 | BAC35196.1 | |
AK137703.1 | 2 | 1568 | BAE23469.1 | |
AK153914.1 | 2 | 1588 | BAE32252.1 | |
BC096491.1 | 2 | 1609 | AAH96491.1 | |
BQ562712.1 | 1 | 306 | ||
JC209414.1 | 2 | 1589 |
Target Gene Details
Entrez Gene ID: | 17281 |
Gene Name: | FYVE and coiled-coil domain containing 1 |
Gene Aliases: |
2810409M01Rik, Mem2, RUFY3, ZFYVE7 |
Location: |
Chr.9:123789500-123851899 on Build GRCm38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fyco1 | NM_001110253.2 | NP_001103723.2 | ||
NM_148925.3 | NP_683727.3 | |||
AJ428065.1 | CAD20987.1 | |||
AK044587.1 | BAE43322.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |