Assay Details
Target Gene Details
Entrez Gene ID: | 13051 |
Gene Name: | chemokine (C-X3-C motif) receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.9:120048683-120068296 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cx3cr1 | NM_009987.4 | 2 | 707 | NP_034117.3 |
AF102269.1 | 2 | 701 | AAC72408.1 | |
AK035902.1 | 2 | 682 | BAC29236.1 | |
AK044557.1 | 2 | 708 | BAC31979.1 | |
AK045634.1 | 2 | 695 | BAC32440.1 | |
AK137388.1 | 2 | 695 | BAE23340.1 | |
AK140010.1 | 2 | 681 | BAE24210.1 | |
BC012653.1 | 2 | 663 | AAH12653.1 |