Assay Details
Target Gene Details
Entrez Gene ID: | 12354 |
Gene Name: | carbonic anhydrase 7 |
Gene Aliases: |
AV343731, Ca7 |
Location: |
Chr.8:104534700-104550347 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car7 | NM_001301164.1 | 7 | 1281 | NP_001288093.1 |
NM_001301165.1 | 7 | 849 | NP_001288094.1 | |
NM_053070.3 | 7 | 876 | NP_444300.1 | |
AF291660.1 | AAG16230.1 | |||
AK090390.1 | 7 | 880 | ||
AK144275.1 | 7 | 850 | BAE25808.1 | |
AY075021.1 | 7 | 877 | AAL78169.1 | |
BC094913.1 | 7 | 864 | AAH94913.1 |