Assay Details
Target Gene Details
Entrez Gene ID: | 13057 |
Gene Name: | cytochrome b-245, alpha polypeptide |
Gene Aliases: |
b558, nmf333, p22-phox, p22phox |
Location: |
Chr.8:122424771-122432965 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyba | NM_001301284.1 | 5 | 468 | NP_001288213.1 |
NM_007806.3 | 6 | 552 | NP_031832.2 | |
AK018713.1 | 5 | 437 | BAB31361.1 | |
AK021200.1 | 6 | 540 | BAB32327.1 | |
AK078854.1 | 6 | 523 | BAC37421.1 | |
AK131580.1 | 6 | 521 | ||
AK152414.1 | 6 | 543 | BAE31199.1 | |
AK171748.1 | 6 | 521 | BAE42648.1 | |
AK190653.1 | ||||
BC026791.1 | 6 | 503 | AAH26791.1 | |
CF738398.1 | 6 | 535 | ||
EU791539.1 | 6 | 482 | ||
M31775.1 | 6 | 475 | AAA37513.1 |