Assay Details
Target Gene Details
Entrez Gene ID: | 73032 |
Gene Name: | tetratricopeptide repeat domain 9B |
Gene Aliases: |
2900074C18Rik |
Location: |
Chr.7:27653924-27656207 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc9b | NM_028417.1 | 1 | 211 | NP_082693.1 |
AK013779.1 | 1 | 214 | BAB28991.1 | |
BC089566.1 | 1 | 211 | AAH89566.1 |
More Information
Set Membership: |
Intragenic Exonic |