Assay Details
Target Gene Details
Entrez Gene ID: | 12700 |
Gene Name: | cytokine inducible SH2-containing protein |
Gene Aliases: |
AI385595, CIS1, Cis, F17, F23 |
Location: |
Chr.9:107296685-107301962 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cish | NM_009895.3 | 3 | 1366 | NP_034025.1 |
AK077338.1 | 3 | 1366 | BAC36758.1 | |
AK170116.1 | 2 | 941 | BAE41574.1 | |
AK170410.1 | 2 | 944 | BAE41776.1 | |
AK170554.1 | 3 | 1167 | BAE41877.1 | |
AK170877.1 | 3 | 1162 | BAE42087.1 | |
AK171431.1 | 3 | 1167 | BAE42448.1 | |
BC003783.1 | 3 | 1170 | AAH03783.1 | |
BC022178.1 | 3 | 1131 | AAH22178.1 | |
D31943.1 | 3 | 1202 | BAA06713.1 |
More Information
Set Membership: |
Intragenic Exonic |