Assay Details
Target Gene Details
Entrez Gene ID: | 12700 |
Gene Name: | cytokine inducible SH2-containing protein |
Gene Aliases: |
AI385595, CIS1, Cis, F17, F23 |
Location: |
Chr.9:107296685-107301962 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cish | NM_009895.3 | 1 | 116 | NP_034025.1 |
AK077338.1 | 1 | 116 | BAC36758.1 |
More Information
Set Membership: |
Intragenic Exonic |