Assay Details
Target Gene Details
Entrez Gene ID: | 67109 |
Gene Name: | zinc finger protein 787 |
Gene Aliases: |
2210018M03Rik, Znf787 |
Location: |
Chr.7:6131489-6159564 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp787 | NM_001013012.1 | 3 | 1619 | NP_001013030.1 |
AK077458.1 | 3 | 1586 | BAC36810.1 | |
AK159396.1 | 2 | 1444 | ||
BC137753.1 |
More Information
Set Membership: |
Intragenic Exonic |