Assay Details
Target Gene Details
Entrez Gene ID: | 73381 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 2A |
Gene Aliases: |
1700001K04Rik, 1700041N15Rik, 1700063K20Rik, AI449770, ARR19, C32, CKLF3, CKLF4, CKLF5, Cklf, Cklf1, Cklfsf2-1b, Cklfsf2a, UCK-1 |
Location: |
Chr.8:104281042-104293181 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtm2a | NM_027022.4 | 5 | 1016 | NP_081298.1 |
AK005603.1 | 5 | 969 | BAB24146.1 | |
AK161303.1 | 5 | 1010 | ||
AY241871.1 | 5 | 1016 | AAP33493.1 | |
BC049747.1 | 5 | 1009 | AAH49747.1 |
More Information
Set Membership: |
Intragenic Exonic |