Assay Details
Target Gene Details
Entrez Gene ID: | 72361 |
Gene Name: | carboxylesterase 2G |
Gene Aliases: |
2210023G05Rik, Ces2l |
Location: |
Chr.8:104961611-104969537 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ces2g | NM_197999.2 | 5 | 843 | NP_932116.2 |
AK146261.1 | 5 | 787 | ||
BC027185.1 | 5 | 821 | AAH27185.1 |
More Information
Set Membership: |
Intragenic Exonic |