Assay Details
Target Gene Details
Entrez Gene ID: | 66113 |
Gene Name: | apolipoprotein A-V |
Gene Aliases: |
1300007O05Rik, Apoav, RAP3 |
Location: |
Chr.9:46268608-46271919 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoa5 | NM_080434.3 | 3 | 1957 | NP_536682.2 |
AK050280.1 | 3 | 1958 | BAC34163.1 |
More Information
Set Membership: |
Intragenic Exonic |