Assay Details
Target Gene Details
Entrez Gene ID: | 11808 |
Gene Name: | apolipoprotein A-IV |
Gene Aliases: |
Apoa-4 |
Location: |
Chr.9:46240844-46243458 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoa4 | NM_007468.2 | 3 | 881 | NP_031494.2 |
AK004856.1 | 3 | 664 | BAB23620.1 | |
AK161535.1 | 3 | 664 | BAE36447.1 | |
AK168687.1 | 3 | 665 | BAE40533.1 | |
AK169277.1 | 3 | 751 | ||
BC010769.1 | 3 | 674 | AAH10769.1 | |
BC050149.1 | 3 | 639 | AAH50149.1 | |
M64248.1 | 3 | 663 | AAA37214.1 | |
M64249.1 | 3 | 663 | AAA37215.1 | |
M64250.1 | 3 | 663 | AAA37216.1 |
More Information
Set Membership: |
Intragenic Exonic |