Assay Details
Target Gene Details
Entrez Gene ID: | 235606 |
Gene Name: | acylpeptide hydrolase |
Gene Aliases: |
- |
Location: |
Chr.9:108085413-108094548 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apeh | NM_146226.2 | 22 | 2212 | NP_666338.2 |
AI845932.1 | 1 | 196 | ||
AK149671.1 | 22 | 2217 | ||
AK149713.1 | 22 | 2213 | ||
AK204166.1 | 1 | 87 | ||
BB737329.1 | 3 | 228 | ||
BC025494.1 | 22 | 2149 | AAH25494.1 | |
BC026594.1 | 17 | 1745 | AAH26594.1 | |
BC034199.1 | 7 | 715 | AAH34199.1 |
More Information
Set Membership: |
Intragenic Exonic |