Assay Details
Target Gene Details
Entrez Gene ID: | 232976 |
Gene Name: | zinc finger protein 574 |
Gene Aliases: |
A630056B21Rik, Znf574 |
Location: |
Chr.7:25075684-25083492 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp574 | NM_001168506.1 | 2 | 1158 | NP_001161978.1 |
NM_175477.4 | 2 | 1213 | NP_780686.1 | |
AK042074.1 | 2 | 1176 | BAC31155.1 | |
AK053744.1 | 2 | 1099 | BAC35502.1 | |
AK143997.1 | 2 | 1159 | BAE25656.1 | |
AK170243.1 | 2 | 1064 | BAE41657.1 | |
AK198096.1 | ||||
BC059044.1 | 2 | 1084 | AAH59044.1 |
More Information
Set Membership: |
Intragenic Exonic |