Assay Details
Target Gene Details
Entrez Gene ID: | 11739 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 4 |
Gene Aliases: |
AU019225, Ant1 |
Location: |
Chr.8:46207339-46211009 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a4 | NM_007450.4 | 2 | 283 | NP_031476.3 |
AK051172.1 | 2 | 306 | ||
AK078077.1 | 2 | 305 | BAC37117.1 | |
AK176986.1 | 2 | 123 | ||
AK197947.1 | 2 | 123 | ||
AK199193.1 | 2 | 123 | ||
BC003791.1 | 2 | 283 | AAH03791.1 | |
BC026925.1 | 2 | 294 | AAH26925.1 | |
U27315.1 | 2 | 282 | AAC52837.1 | |
X74510.1 | 2 | 289 | CAA52616.1 |
More Information
Set Membership: |
Intragenic Exonic |