Assay Details
Target Gene Details
Entrez Gene ID: | 53896 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 10 |
Gene Aliases: |
AL024237, Asc-1, D7Bwg0847e |
Location: |
Chr.7:35186357-35201114 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a10 | NM_017394.4 | 1 | 86 | NP_059090.3 |
AB026688.1 | BAA93617.1 | |||
AK031867.1 | 1 | 54 | BAC27582.1 | |
BC054765.1 | 1 | 29 | AAH54765.1 |
More Information
Set Membership: |
Intragenic Exonic |