Assay Details
Target Gene Details
Entrez Gene ID: | 14132 |
Gene Name: | Fc receptor, IgG, alpha chain transporter |
Gene Aliases: |
FcRn |
Location: |
Chr.7:45092992-45103846 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fcgrt | NM_010189.3 | 7 | 1686 | NP_034319.2 |
AK146933.1 | 7 | 1651 | ||
BC003786.1 | AAH03786.1 | |||
D37874.1 | BAA07111.1 | |||
L17022.1 | AAA16904.1 |
More Information
Set Membership: |
Intragenic Exonic |