Assay Details
Target Gene Details
Entrez Gene ID: | 243967 |
Gene Name: | netrin 5 |
Gene Aliases: |
Gm484 |
Location: |
Chr.7:45684022-45694556 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ntn5 | NM_001033356.3 | 6 | 1877 | NP_001028528.2 |
NM_001289692.1 | 7 | 1936 | NP_001276621.1 | |
NM_001289693.1 | 7 | 1670 | NP_001276622.1 | |
AK042348.1 | 7 | 1672 | ||
AK142898.1 | 6 | 1485 | BAE25222.1 | |
BC150901.1 | ||||
BX639313.1 | 1 | 95 |
More Information
Set Membership: |
Intragenic Exonic |