Assay Details
Target Gene Details
Entrez Gene ID: | 17304 |
Gene Name: | milk fat globule-EGF factor 8 protein |
Gene Aliases: |
AA408458, AI325141, MFG-E8, MP47, Mfgm, P47, SED1, lactadherin |
Location: |
Chr.7:79133768-79149060 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mfge8 | NM_001045489.1 | 9 | 1371 | NP_001038954.1 |
NM_008594.2 | 10 | 1482 | NP_032620.2 | |
AB021130.2 | BAA35180.1 | |||
AB025280.1 | BAA76386.1 | |||
AK087555.1 | 9 | 1361 | ||
AK089211.1 | 9 | 1337 | BAC40794.1 | |
AK145183.1 | 9 | 1337 | ||
AK148371.1 | 9 | 1360 | BAE28512.1 | |
AK152088.1 | 10 | 1471 | BAE30938.1 | |
AK155047.1 | 10 | 1480 | ||
AK169997.1 | 9 | 1335 | BAE41503.1 | |
AK170935.1 | 9 | 1335 | BAE42127.1 | |
AK171143.1 | 10 | 1446 | BAE42274.1 | |
AK195377.1 | ||||
AK196442.1 | ||||
AK207636.1 | ||||
BC003892.1 | 9 | 1315 | AAH03892.1 | |
BC003904.1 | 9 | 1310 | AAH03904.1 | |
M38337.1 | 10 | 1434 | AAA39534.1 | |
Y11684.2 | CAA72380.2 |
More Information
Set Membership: |
Intragenic Exonic |