Assay Details
Target Gene Details
Entrez Gene ID: | 171504 |
Gene Name: | apolipoprotein B receptor |
Gene Aliases: |
Apob-48r, Apob48r |
Location: |
Chr.7:126584912-126589125 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apobr | NM_138310.1 | 3 | 2536 | NP_612183.1 |
AF141335.1 | 3 | 2536 | AAL54862.1 | |
AK156422.1 | 3 | 2604 | ||
BC030718.1 | 3 | 2544 | AAH30718.1 |