Assay Details
Target Gene Details
Entrez Gene ID: | 69748 |
Gene Name: | aldehyde dehydrogenase 16 family, member A1 |
Gene Aliases: |
2410004H02Rik, AI452234, mFLJ00209 |
Location: |
Chr.7:45141833-45154538 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh16a1 | NM_145954.1 | 17 | 2391 | NP_666066.1 |
AK010387.1 | 4 | 494 | BAB26902.2 | |
AK030673.1 | 17 | 2384 | BAC27074.1 | |
AK144995.1 | 17 | 2398 | BAE26175.1 | |
AK151691.1 | 17 | 2391 | BAE30615.1 | |
AK153129.1 | 10 | 1298 | BAE31742.1 | |
AK161198.1 | 17 | 2429 | BAE36234.1 | |
AK170276.1 | 17 | 2430 | BAE41680.1 | |
AK220200.1 | 17 | 2367 | BAD90125.1 | |
BC013548.1 | 17 | 2391 | AAH13548.2 | |
BC030467.1 | 4 | 599 | AAH30467.1 |
More Information
Set Membership: |
Intragenic Exonic |