Assay Details
Target Gene Details
Entrez Gene ID: | 70080 |
Gene Name: | immunoglobulin superfamily, member 23 |
Gene Aliases: |
2210010C17Rik |
Location: |
Chr.7:19937305-19950765 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf23 | NM_027308.2 | 3 | 558 | NP_081584.1 |
AK008696.1 | 3 | 560 | BAB25838.1 | |
AK033677.1 | 3 | 559 | BAC28424.1 | |
AK088927.1 | 4 | 514 | BAC40656.1 | |
BC139455.1 | 3 | 406 | ||
BC139458.1 | 3 | 406 |