Assay Details
Target Gene Details
Entrez Gene ID: | 70080 |
Gene Name: | immunoglobulin superfamily, member 23 |
Gene Aliases: |
2210010C17Rik |
Location: |
Chr.7:19937305-19950765 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf23 | NM_027308.2 | 1 | 55 | NP_081584.1 |
AK008696.1 | 1 | 57 | BAB25838.1 | |
AK033677.1 | 1 | 56 | BAC28424.1 |