Assay Details
Target Gene Details
Entrez Gene ID: | 24030 |
Gene Name: | mitochondrial ribosomal protein S12 |
Gene Aliases: |
AI327385, Rpms12, rps12 |
Location: |
Chr.7:28739641-28741890 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrps12 | NM_011885.4 | 3 | 480 | NP_036015.1 |
AK146730.1 | 3 | 502 | ||
BC012679.1 | 3 | 516 | AAH12679.1 | |
BC027522.1 | 3 | 485 | AAH27522.1 | |
Y11682.1 | 3 | 479 | CAA72378.1 |
More Information
Set Membership: |
Intragenic Exonic |