Assay Details
Target Gene Details
Entrez Gene ID: | 654804 |
Gene Name: | RIKEN cDNA 4732471J01 gene |
Gene Aliases: |
- |
Location: |
Chr.7:25376819-25566417 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4732471J01Rik | NR_073027.1 | |||
NR_073028.1 | ||||
AK043644.1 | ||||
AK076372.1 | ||||
AK078204.1 | BAC37171.1 | |||
BB613302.1 |
Target Gene Details
Entrez Gene ID: | 16890 |
Gene Name: | lipase, hormone sensitive |
Gene Aliases: |
4933403G17Rik, HSL |
Location: |
Chr.7:25379527-25398704 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lipe | NM_001039507.2 | 10 | 2357 | NP_001034596.1 |
NM_010719.5 | 10 | 2793 | NP_034849.2 | |
AK029984.1 | 10 | 2796 | BAC26716.1 | |
AK169858.1 | 10 | 2352 | BAE41414.1 | |
BC021642.1 | 10 | 2353 | AAH21642.1 | |
U08188.1 | 9 | 2878 | AAC52163.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |