Assay Details
Target Gene Details
Entrez Gene ID: | 13360 |
Gene Name: | 7-dehydrocholesterol reductase |
Gene Aliases: |
AA409147 |
Location: |
Chr.7:143823167-143848410 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhcr7 | NM_007856.2 | 9 | 1683 | NP_031882.1 |
AF057368.1 | 8 | 1215 | AAC40164.1 | |
AK146811.1 | 8 | 1206 | BAE27451.1 | |
AK190502.1 | ||||
BC006854.1 | 7 | 1197 | AAH06854.1 | |
CT010277.1 | 7 | 1176 | CAJ18485.1 |
More Information
Set Membership: |
Intragenic Exonic |