Assay Details
Target Gene Details
Entrez Gene ID: | 93695 |
Gene Name: | glycoprotein (transmembrane) nmb |
Gene Aliases: |
DC-HIL, Dchil, ipd |
Location: |
Chr.6:49036518-49058182 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpnmb | NM_053110.4 | 11 | 2119 | NP_444340.3 |
AF322054.1 | 11 | 1957 | AAK14240.1 | |
AJ251685.1 | 11 | 2004 | CAB65272.1 | |
AK044764.1 | 11 | 2013 | BAC32074.1 | |
AK076347.1 | 11 | 1988 | BAC36306.1 | |
AK149740.1 | ||||
AK150739.1 | 11 | 1988 | BAE29813.1 | |
AK154617.1 | 11 | 2023 | BAE32716.1 | |
AK159165.1 | 11 | 2018 | BAE34867.1 | |
AK159270.1 | 11 | 2026 | BAE34949.1 | |
AK159747.1 | 11 | 1988 | BAE35339.1 | |
AK159908.1 | 11 | 1993 | ||
AK170506.1 | 11 | 2018 | BAE41844.1 | |
AK171347.1 | 11 | 1994 | BAE42404.1 | |
AK171441.1 | 11 | 1988 | BAE42454.1 | |
AK171465.1 | 11 | 1988 | BAE42471.1 | |
AK171474.1 | 11 | 1988 | BAE42479.1 | |
AK171517.1 | 11 | 2106 | BAE42501.1 | |
AK171608.1 | 11 | 1988 | BAE42560.1 | |
AK171618.1 | 11 | 1988 | BAE42567.1 | |
BC026375.1 | 11 | 1993 | AAH26375.1 |
More Information
Set Membership: |
Intragenic Exonic |