Assay Details
Target Gene Details
Entrez Gene ID: | 12183 |
Gene Name: | 2,3-bisphosphoglycerate mutase |
Gene Aliases: |
AI323730, AL022789, C86192 |
Location: |
Chr.6:34476254-34505610 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bpgm | NM_007563.4 | 3 | 1848 | NP_031589.1 |
AK078119.1 | 3 | 1849 | BAC37133.1 |
More Information
Set Membership: |
Intragenic Exonic |