Assay Details
Target Gene Details
Entrez Gene ID: | 18861 |
Gene Name: | postmeiotic segregation increased 2 (S. cerevisiae) |
Gene Aliases: |
AW555130, Pmsl2 |
Location: |
Chr.5:143909974-143985719 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pms2 | NM_008886.2 | 11 | 1766 | NP_032912.2 |
AK034866.1 | 11 | 1742 | ||
AK083049.1 | 11 | 1723 | ||
AK146365.1 | 11 | 1779 | BAE27115.1 | |
BC139774.1 | ||||
BC141287.1 | 11 | 1742 |
More Information
Set Membership: |
Intragenic Exonic |