Assay Details
Target Gene Details
Entrez Gene ID: | 320135 |
Gene Name: | cDNA sequence BC049715 |
Gene Aliases: |
- |
Location: |
Chr.6:136828160-136841920 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BC049715 | NM_178776.3 | 3 | 593 | NP_848891.2 |
AK044622.1 | 2 | 1808 | BAC32007.1 | |
BC049715.1 | 3 | 593 | AAH49715.1 |