Assay Details
Target Gene Details
Entrez Gene ID: | 106840 |
Gene Name: | unc-119 homolog B (C. elegans) |
Gene Aliases: |
AA407659, AI414892 |
Location: |
Chr.5:115122566-115134975 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Unc119b | NM_175352.4 | 1 | 59 | NP_780561.1 |
AK045128.1 | BAC32235.1 | |||
AK082614.1 | 1 | 61 | BAC38549.1 | |
BC112388.1 | ||||
BC115529.2 |
More Information
Set Membership: |
Intragenic Exonic |