Assay Details
Target Gene Details
Entrez Gene ID: | 257635 |
Gene Name: | serine dehydratase-like |
Gene Aliases: |
4432411H13Rik, AI504310, SDH1, SDS-RS1, TDH |
Location: |
Chr.5:120458193-120472891 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdsl | NM_133902.2 | 8 | 1156 | NP_598663.2 |
AF328927.1 | 9 | 1423 | AAL56988.1 | |
AK014487.1 | ||||
BC022601.1 | 8 | 1156 | AAH22601.1 |
More Information
Set Membership: |
Intragenic Exonic |