Assay Details
Target Gene Details
Entrez Gene ID: | 22697 |
Gene Name: | zinc finger and SCAN domain containing 21 |
Gene Aliases: |
AI326272, CTfin51, RU49, Zfp-38, Zfp38, Zipro1 |
Location: |
Chr.5:138116899-138134265 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zscan21 | NM_001044703.2 | 2 | 211 | NP_001038168.1 |
NM_001044704.2 | 3 | 277 | NP_001038169.1 | |
NM_001044705.2 | 2 | 192 | NP_001038170.1 | |
NM_011757.3 | 3 | 296 | NP_035887.2 | |
AK146776.1 | 3 | 270 | ||
AK146875.1 | 3 | 257 | BAE27498.1 | |
BC010591.1 | 2 | 139 | AAH10591.1 | |
BU513148.1 | 2 | 153 | ||
BY034027.1 | 3 | 281 | ||
BY293243.1 | 3 | 278 | ||
BY309906.1 | 2 | 179 | ||
CF915830.1 | 3 | 221 | ||
D10630.1 | 2 | 130 | BAA01480.1 | |
DV651309.1 | 3 | 309 | ||
U41671.1 | 3 | 227 | AAB03786.1 | |
X63747.1 | 2 | 987 | CAA45280.1 |
More Information
Set Membership: |
Intragenic Exonic |