Assay Details
Target Gene Details
Entrez Gene ID: | 20751 |
Gene Name: | sepiapterin reductase |
Gene Aliases: |
AA409688, ENSMUSG00000071345, Gm10328 |
Location: |
Chr.6:85133680-85137764 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spr | NM_011467.2 | 1 | 273 | NP_035597.2 |
AK004941.1 | 1 | 265 | ||
BC010400.1 | 1 | 270 | AAH10400.1 | |
BC054423.1 | 1 | 257 | AAH54423.1 | |
BY113096.1 | 1 | 274 | ||
CK620382.1 | 1 | 241 | ||
S77493.1 | 1 | 275 | AAB33611.1 |
More Information
Set Membership: |
Intragenic Exonic |