Assay Details
Target Gene Details
Entrez Gene ID: | 18542 |
Gene Name: | procollagen C-endopeptidase enhancer protein |
Gene Aliases: |
Astt-2, Astt2, P14, PCPE-1 |
Location: |
Chr.5:137605105-137611899 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcolce | NM_008788.2 | 6 | 855 | NP_032814.2 |
AB008548.1 | 6 | 779 | BAA23280.1 | |
AK142615.1 | 6 | 784 | BAE25130.1 | |
AK144392.1 | 6 | 856 | BAE25865.1 | |
AK146829.1 | 6 | 856 | BAE27464.1 | |
AK159430.1 | 6 | 843 | BAE35077.1 | |
AK159940.1 | 6 | 848 | ||
AK162524.1 | 6 | 859 | ||
CX210896.1 | 4 | 537 | ||
X57337.1 | 6 | 822 | CAA40612.1 |
More Information
Set Membership: |
Intragenic Exonic |