Assay Details
Target Gene Details
Entrez Gene ID: | 18979 |
Gene Name: | paraoxonase 1 |
Gene Aliases: |
Pon |
Location: |
Chr.6:5168090-5193987 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pon1 | NM_011134.3 | 9 | 1241 | NP_035264.2 |
AI194741.1 | 1 | 314 | ||
BC012706.1 | 9 | 1114 | AAH12706.1 | |
U32684.1 | 9 | 1123 | AAC52496.1 | |
U72636.1 | 9 | 1123 | AAB17394.1 |