Assay Details
Target Gene Details
Entrez Gene ID: | 75387 |
Gene Name: | sirtuin 4 |
Gene Aliases: |
4930596O17Rik |
Location: |
Chr.5:115478006-115490050 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sirt4 | NM_001167691.1 | 2 | 659 | NP_001161163.1 |
NM_133760.1 | 2 | 567 | NP_598521.1 | |
AK016400.1 | 2 | 659 | ||
BC022653.1 | 2 | 435 | AAH22653.1 |