Assay Details
Target Gene Details
Entrez Gene ID: | 14911 |
Gene Name: | THUMP domain containing 3 |
Gene Aliases: |
AW556087, Gt(ROSA)26asSor, Gtrosa26as |
Location: |
Chr.6:113046252-113068278 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Thumpd3 | NM_008188.2 | 4 | 513 | NP_032214.1 |
AK017246.1 | 4 | 526 | BAB30650.1 | |
AK142140.1 | 4 | 532 | BAE24945.1 | |
BC002024.1 | 4 | 515 | AAH02024.1 | |
BC012688.1 | 3 | 461 | AAH12688.1 | |
BC046800.1 | 4 | 513 | AAH46800.1 | |
U83176.1 | 4 | 507 | AAC60384.1 |
More Information
Set Membership: |
Intragenic Exonic |