Assay Details
Target Gene Details
Entrez Gene ID: | 19713 |
Gene Name: | ret proto-oncogene |
Gene Aliases: |
PTC, RET51, RET9, c-Ret |
Location: |
Chr.6:118151748-118197744 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ret | NM_001080780.1 | 19 | 6210 | NP_001074249.1 |
NM_009050.2 | 20 | 4933 | NP_033076.2 | |
AK162564.1 | 1 | 1035 |
More Information
Set Membership: |
Intragenic Exonic |