Assay Details
Target Gene Details
Entrez Gene ID: | 18096 |
Gene Name: | NK6 homeobox 1 |
Gene Aliases: |
NKX6A, Nkx6.1 |
Location: |
Chr.5:101659184-101664711 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nkx6-1 | NM_144955.2 | 1 | 767 | NP_659204.1 |
AF357883.1 | 1 | 767 | AAK37567.1 | |
BC138019.1 | 1 | 673 | ||
BC138020.1 | 1 | 673 |
More Information
Set Membership: |
Intragenic Exonic |