Assay Details
Target Gene Details
Entrez Gene ID: | 231997 |
Gene Name: | FK506 binding protein 14 |
Gene Aliases: |
BC029109, FKBP-14, FKBP22 |
Location: |
Chr.6:54577605-54593698 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp14 | NM_153573.1 | 1 | 237 | NP_705801.1 |
AK030620.1 | 1 | 287 | ||
AK044876.1 | 1 | 273 | ||
AK081692.1 | 1 | 264 | BAC38295.1 | |
BC029109.1 | 1 | 237 | AAH29109.1 | |
BC034313.1 | 1 | 262 |
More Information
Set Membership: |
Intragenic Exonic |