Assay Details
Target Gene Details
Entrez Gene ID: | 18028 |
Gene Name: | nuclear factor I/B |
Gene Aliases: |
6720429L07Rik, CTF, E030026I10Rik, NF-I/B, NF1-B, NFI-B |
Location: |
Chr.4:82290173-82505779 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfib | NM_001113209.2 | 12 | 4820 | NP_001106680.1 |
NM_001113210.2 | 12 | 4793 | NP_001106681.1 | |
NM_001286127.1 | 11 | 4704 | NP_001273056.1 | |
NM_001286131.1 | 10 | 4571 | NP_001273060.1 | |
NM_008687.6 | 9 | 4482 | NP_032713.3 | |
AK049772.1 | 1 | 868 | ||
AK053181.1 | 1 | 868 | BAC35299.1 | |
AK081555.1 | 1 | 868 |
More Information
Set Membership: |
Intragenic Exonic |